ContactUsLinkComponent
μPAC™ ESI-MS Accessories
μPAC™ ESI-MS Accessories
Catalog # HYPEEMI-EASYSPRAYB
μPAC™ ESI-MS Accessories
Catalog # HYPEEMI-EASYSPRAYB
Specifications
  • For:
    EASY-Spray™ source
  • Description:
    μPAC™ compatible EASY-spray emitter
  • Pk:
    1
Specifications
About this item
ESI-MS accessories are used to facilitate reproducible, low dispersion connections between μPAC HPLC columns and low-flow mass spectrometer ion sources.
The μPAC EASY-spray emitter is equipped with a stainless steel 50 μm through-bore union. Achieve high resolution separations in low-flow applications when using this emitter to connect a μPAC HPLC column and a transfer line with a EASY-Spray™ source.
The μPAC EASY-spray emitter is designed for easy connection to an EASY-Spray MS source. It contains a pre-assembled EASY-Spray emitter with a transfer line and grounding union.