ContactUsLinkComponent
Experimentation kits, analysis and comparision of mammalian cell types
Experimentation kits, analysis and comparision of mammalian cell types
Catalog # EDVO986
Supplier:  EDVOTEK
undefined
Experimentation kits, analysis and comparision of mammalian cell types
Catalog # EDVO986
Supplier:  EDVOTEK
Specifications
  • Description:
    Analysis and Comparision of Mammalian Cell Types
  • Pk:
    1
Specifications
About this item
Observe the differences between mammalian cell types and how these cells funciton. Cells are fixed on microscope slides and students stain the cells on the slide to view morphological characteristics of the cell types. These cells are very safe for classroom use.
  • For 6 lab groups
  • Complete in 35 minutes
  • Requires a microscope with 400× magnification
Delivery: Kit includes instructions, multispot slides (4 cell types each), eosin and methylene blue stain, mounting medium, cover slips, transfer pipettes, and immersion troughs.