Orders LinkContactUsLinkComponent
Western blotting blocking reagent, Amersham™ ECL™
Amersham ECL™ blocking agent, 40 g
Catalog # RPN2125
Supplier:   Cytiva
Western blotting blocking reagent, Amersham™ ECL™
Catalog # RPN2125
Supplier:  Cytiva
CAS Number:  
Western blotting blocking reagent, Amersham™ ECL™
Western blotting blocking reagent, Amersham™ ECL™
Amersham ECL™ blocking agent, 40 g
CAS Number:  
Price
91,90 €
Log in for availability
1 * 40 g
Product Details & Documents
Amersham ECL Blocking Agent is for blocking nitrocellulose and PVDF membranes to be used with Amersham ECL, Amersham ECL Prime and Amersham ECL Select Western Blotting Detection Reagents
Can be used with the Amersham ECL Western blotting detection family. This blocking agent is intended for blocking nitrocellulose and PVDF membranes used with ECL and ECL Plus Western Blotting Detection Reagents.
Delivery information: Each pack contains 40 g of membrane blocking agent, sufficient for at least 20 miniblots.
Specifications
Specifications
  • Pk:
    40 g
  • Description:
    Amersham ECL™ blocking agent