Order Entry
Austria
ContactUsLinkComponent
Mobile Box for Hazardous Materials
Mobile Box for Hazardous Materials
Catalog # CEMG11455
Supplier:  CEMO
Mobile Box for Hazardous Materials
Catalog # CEMG11455
Supplier:  CEMO

Specifications

  • Capacity:
    170 L
  • Description:
    MOBILE box with green lid for hazardous materials,
    certified, Weight: 12 kg
  • W×D×H:
    400×600×880 mm
  • Pk:
    1

Specifications

About this item

The robust design enables clean and safe storage of hazardous solid goods belonging to packing groups II or III and is also certified for transport according to ADR, RID and IMDG codes. The container (grey), with a capacity of 170 L, and lid (coloured) are easy to handle, even when the container is full.

  • Lockable mechanism with metal bracket and locking spring
  • Lid with PU seal, fire-classified, light and air-tight for moisture-sensitive material
  • Up to 8 containers on a euro pallet, two layer stackable
  • It can be used as collection container for waste and materials such as oil binding agents, paint residues, etc
  • It can be used as collection and transport container for recycling, cleaning cloths, etc
  • It can be used as collection container for flammable and polluting substances

The specially shaped lid enables storage in 2-layers. Up to 8 boxes can be safely stacked on a Euro pallet. The beading in the container wall enables emptying with a rotary forklift.