Orders LinkContactUsLinkComponent
Pasteur Pipettes
Pasteur Pipettes
Supplier:   KARTELL
Pasteur Pipettes
Supplier:  KARTELL
Certificates
Pasteur Pipettes
Pasteur Pipettes
3 Options Available Below
Product Details & Documents
Disposable polyethylene pipette with bulb and graduations. The slender tip guarantees precision transfer of the sample.
  • Device for professional use in laboratory analysis
  • Slender tip guarantees precision transfer of the sample
  • Overall pipette length: 150 mm
Non sterile pipettes are available in multi-packs. Sterile pipettes are individually wrapped.
Product Family Options
Product Information
  • Drops per ml
    Bulb draw
    Stem Ø
    Sterile
    Availability
    Price
  • 28
    3,5 ml
    2,5 mm
    -
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 256,00 €
    256,00 €
    1 * 6 ST
       256,00 €
  • 23
    3,5 ml
    5 mm
    -
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 228,00 €
    228,00 €
    1 * 6 ST
       228,00 €
  • 21
    3,5 ml
    7,8 mm
    +
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 107,00 €
    107,00 €
    1 * 500 ST
       107,00 €