ContactUsLinkComponent
Sterile Pipette Filter Tips in Rack
Artikelnr. KART0963500
Lieferant:  KARTELL
undefined
Sterile Pipette Filter Tips in Rack
Artikelnr. KART0963500
Lieferant:  KARTELL
Spezifikationen
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    2 - 100 µl
  • Length:
    50 mm
  • Tip style:
    Low retention; graduated
  • Tip colour:
    Natural
  • For use with:
    Kartell® PL100, Gilson®, Eppendorf ®, Biohit®, Socorex®, HTL®, Nichiryo®, Finnpipette®, Thermo®
  • Filter:
    +
  • Pk:
    10
Spezifikationen
Über diesen Artikel
These tips are made of high quality, 100% pure, virgin PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • Sterilised by gamma radiation
  • Designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology
  • Hydrophobic pe filters act as a barrier against aerosol contamination, preventing cross-contamination of samples and micropipettes
  • Tip orifices are sized to completely empty the tip in the event of over-aspiration
Made of PP and including a filter, these tips are DNase, RNase and DNA free, ATP and metal free, cadmiuim free, apyrogenic and PCR inhibitor free.