ContactUsLinkComponent
Compression Moulded Closure for Narrow Neck Bottle Series 310
Compression Moulded Closure for Narrow Neck Bottle Series 310
Catalog # KAUT2000971704
Supplier:  Kautex
undefined
Compression Moulded Closure for Narrow Neck Bottle Series 310
Catalog # KAUT2000971704
Supplier:  Kautex
Specifications
  • Thread:
    25 mm
  • Colour:
    White
  • Material:
    Melamine
  • Pk:
    100
  • For bottles:
    250 - 1000 ml
Specifications
About this item
Compression moulded closure is made from melamine, and features a PE-foam insert.
  • Leakproof