CRS SPure™ H₂O/HC Filters
The SPure H₂O/HC Filter removes moisture, oil, dust, and hydrocarbons (C5 and heavier) from dry air and inert gases.
The cobalt-free moisture indicator signals breakthrough at 5% relative humidity and changes from green to blue.
SPure Filters have the option of standard compression fittings and quick connect fittings in ¹/₄ and ¹/₈" brass and stainless steel, or metal push-to-connect fittings in ¹/₄" and 6 mm for plastic tubing and non-carrier gas lines.
Frequently Bought Together