ContactUsLinkComponent
SPE Plate, 96-well, NBE TRACE-N20
  TCAT611-0051NBE
 :  Tecan
SPE Plate, 96-well, NBE TRACE-N20
  TCAT611-0051NBE
 :  Tecan
  • Description:
    Plate NBE TRACE-N20, 1ml, 20 µm
  • Environmentally Preferable:
  • Phase:
    TRACE-N20
  • Filling:
    5 mg
  • Pk:
    1
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
  • Reversed-phase
  • Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.