Order Entry
Austria
ContactUsLinkComponent
SPE Plate, 96-well, NBE PSAX
SPE Plate, 96-well, NBE PSAX
  TCAT615-0051NBE
 :  Tecan
SPE Plate, 96-well, NBE PSAX
  TCAT615-0051NBE
 :  Tecan

 

  • No. of wells:
    96
  • Description:
    Plate NBE PSAX 1 ml; 15 µm
  • Phase:
    PSAX
  • Filling:
    5 mg
  • Pk:
    1

 

 

Mixed mode reverse-phase with an anion exchanger for acidic compounds.

  • Ion-exchange
  • Liquid handler compatible

A 17 µm polymer micro particulate with strong anionic exchange chemistry providing large contat zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for acidic target compounds.