SPE Plate, 96-well, NBE OFX
TCAT633-0031NBE
: Tecan
- No. of wells:96
- Description:Plate NBE OFX 1 ml 30 µm
- Environmentally Preferable:
- Phase:OFX
- Filling:3 mg
- Pk:1
A proprietary polymer resin blend of ion-exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.