Atlas® SPE Plates, 96-well, RCH
Supplier: Tecan
Certificates
About this item
A hydrophilic/lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species, also comes in larger particle for more viscous samples. A direct alternative to Waters Oasis® HLB and Phenomenex Strata X.
- Resin chemistry: N-vinylpyrrolidone-divinylbenzene copolymer
- Particle shape: spherical
- pH stability: 1 - 14
Ideal for use in small molecules and pharmaceutical drugs, sample preparation for drug monitoring, drug screening, and steroid analysis workflows.
Frequently Bought Together