Order Entry
Austria
ContactUsLinkComponent
SPE Column, NBE OFX
SPE Column, NBE OFX
Catalog # TCAT633-0031R-NBE
Supplier:  Tecan
SPE Column, NBE OFX
Catalog # TCAT633-0031R-NBE
Supplier:  Tecan

Specifications

  • Particle Size:
    30 µm
  • Capacity:
    1 ml
  • Description:
    Column NBE OFX
  • Bed weight:
    3 mg
  • Pk:
    96

Specifications

About this item

Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.