Order Entry
Austria
ContactUsLinkComponent
SPE Column, NBE OFQC
SPE Column, NBE OFQC
Catalog # TCAT637-0051R-NBE
Supplier:  Tecan
SPE Column, NBE OFQC
Catalog # TCAT637-0051R-NBE
Supplier:  Tecan

Specifications

  • Particle Size:
    30 µm
  • Capacity:
    1 ml
  • Description:
    Column NBE OFQC
  • Bed weight:
    5 mg
  • Phase:
    OFQC
  • Pk:
    96

Specifications

About this item

A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.