Order Entry
Austria
ContactUsLinkComponent
SPE Column, NBE MAESTRO
SPE Column, NBE MAESTRO
Catalog # TCAT613-0051R-NBE
Supplier:  Tecan
SPE Column, NBE MAESTRO
Catalog # TCAT613-0051R-NBE
Supplier:  Tecan

Specifications

  • Particle Size:
    10 µm
  • Capacity:
    1 ml
  • Description:
    Column NBE MAESTRO
  • Bed weight:
    5,0 mg
  • Phase:
    MAESTRO
  • Pk:
    96

Specifications

About this item

Silica-based sorbent with C18 bonding in proprietary coverage.

  • Reversed-phased
  • Liquid handler compatible

Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.