Order Entry
Austria
ContactUsLinkComponent
SPE Column, NBE HPSAX
SPE Column, NBE HPSAX
Catalog # TCAT412-0021R-NBE
Supplier:  Tecan
SPE Column, NBE HPSAX
Catalog # TCAT412-0021R-NBE
Supplier:  Tecan

Specifications

  • Particle Size:
    20 µm
  • Capacity:
    1 ml
  • Description:
    Column NBE HPSAX
  • Bed weight:
    2,5 mg
  • Pk:
    96

Specifications

About this item

Polymeric strong anionic exchange chemistry.

  • Ion-exchange
  • Liquid handler compatible

20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.