Cerex® Atlas® SCX SPE Columns
: Tecan
A hydrophilic/lipophilic balanced strong cationic chemistry offering excellent selectivity to basic and neutral compounds. A direct alternative to Waters Oasis® MCX and Phenomenex Strata XC.
- Mode: Ion-exchange
- Liquid handler compatible
Atlas® is a hydrophilic/lipophilic balanced polymeric sorbent with a N-vinylpyrrolidone-divinyl-ben-zene copolymer. Atlas® SCX which incorporates the Atlas’s copolymer technology along with a strong cationic exchange chemistry to provide a focused clean up when working with basic compounds in tough matrices.
Frequently Bought Together