Order Entry
Austria
ContactUsLinkComponent
TASKI® Jontec Best Neutral Floor Degreaser, Low Foam
TASKI® Jontec Best Neutral Floor Degreaser, Low Foam
Catalog # DIVY7512309
Supplier:  Diversey
TASKI® Jontec Best Neutral Floor Degreaser, Low Foam
Catalog # DIVY7512309
Supplier:  Diversey

Specifications

  • Capacity:
    5 L
  • Description:
    F4e 2×5 L Neutral floor degreaser
  • Seq:
    01
  • Colour:
    Clear
  • Pack type:
    Bottle
  • Pk:
    2 Bottle
  • pH value:
    8,8 pH

Specifications

About this item

Heavy duty, alkaline floor cleaner. Cleans heavy soil, dirt and grease off all water resistant, non protected hard floors. Especially suitable for heavy duty ceramic and porous tiles cleaning.

  • pH-neutral
  • Good emulsifying power
  • Fast acting, low foaming
  • Versatile application
  • Environmentally responsible for waste water disposal
  • Unique, patented Odour Neutralizing Technology (O.N.T.)

Safe to use on all water resistant hard floors. Ideal for scrubber driers. Removes quickly and thoroughly mineral/synthetic oils and grease, forklift tire marks as well as water-soluble dirt. Low foaming for fast solution pick-up and optimal recovery tank use. Also suitable for wet mopping and single disc machine systems. Leaves a pleasant, fresh fragrance. Suitable to be disposed through oil separators. Neutralises malodour molecules and therefore enhances the perfume effect.