ContactUsLinkComponent
96 Well Plate, RCH2 PPBA
96 Well Plate, RCH2 PPBA
Catalog # TCAT30168995
Supplier:  Tecan
96 Well Plate, RCH2 PPBA
Catalog # TCAT30168995
Supplier:  Tecan
Specifications
  • No. of wells:
    96
  • Description:
    96-well RCH Atlas® plate
  • Phase:
    RCH2
  • Filling:
    50 mg
  • Pk:
    1
Specifications
About this item
A special phenyl boronic acid chemistry on polymer base resin for better capacity and solvent use. PBA utilizes an unique covalent interaction ideal for catecholamine analysis.
  • Cerex
  • Reversed-phased
  • Liquid handler compatible
Particle size: 60 µm