Order Entry
Austria
ContactUsLinkComponent
Cerex® OFQC SPE Column
Cerex® OFQC SPE Column
Catalog # TCAT637-0101R
Supplier:  Tecan
Cerex® OFQC SPE Column
Catalog # TCAT637-0101R
Supplier:  Tecan

Specifications

  • Particle Size:
    30 µm
  • Capacity:
    1 ml
  • Description:
    Column Cerex® OFQC
  • Bed weight:
    10 mg
  • Phase:
    OFQC
  • Pk:
    96

Specifications

About this item

A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.