Orders LinkContactUsLinkComponent
Doypack® Paper/PET/CPP Paper Bags
Doypack® Paper/PET/CPP Paper Bags
Supplier:   Flexico
Doypack® Paper/PET/CPP Paper Bags
Supplier:  Flexico
Certificates
Doypack® Paper/PET/CPP Paper Bags
5 Options Available Below
Product Details & Documents
The specific shape of the Doypack® bags allows a good vertical position of the packaging on the shelves. The Paper/PET/CPP laminate provides a good barrier to gases and humidity. The paper layer gives the bag a natural look.
  • Food contact
  • Available in different sizes
  • Ideal for wet and dry content
  • Doypack® packaging
  • Ideal for shelf display
  • Available in stock
The Doypack® packaging has a window to give a better visibility of the content and highlights the features of the product. Ideal for food products (biscuits, confectionery, snacks) this bag is reclosable and reusable. All Doypack® Stand Up pouches in stock are equipped with a Clicky® closure system, with tactile & sound effects for an intuitive and sensory closure.
Product Family Options
Product Information
  • Capacity
    Material
    Thickness
    W×L
    Availability
    Price
  • 129-3901
    Doypack® Paper/PET/CPP bags, 2×25 mm bottom gusset
    Doypack® Paper/PET/CPP bags, 2×25 mm bottom gusset
    100 ml
    PET/CPP
    PAPER40/PET12/CPP60
    85×125 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 391,00 €
    391,00 €
    1 * 500 ST
       391,00 €
  • 129-3902
    Doypack® Paper/PET/CPP bags, 2×32 mm bottom gusset
    Doypack® Paper/PET/CPP bags, 2×32 mm bottom gusset
    250 ml
    PET/CPP
    PAPER40/PET12/CPP60
    110×160 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 437,00 €
    437,00 €
    1 * 500 ST
       437,00 €
  • 129-3903
    Doypack® Paper/PET/CPP bags, 2×35 mm bottom gusset
    Doypack® Paper/PET/CPP bags, 2×35 mm bottom gusset
    500 ml
    PET/CPP
    PAPER40/PET12/CPP60
    130×190 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 481,00 €
    481,00 €
    1 * 500 ST
       481,00 €
  • 129-3904
    Doypack® Paper/PET/CPP bags, 2×40 mm bottom gusset
    Doypack® Paper/PET/CPP bags, 2×40 mm bottom gusset
    750 ml
    PET/CPP
    PAPER40/PET12/CPP60
    160×230 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 546,00 €
    546,00 €
    1 * 500 ST
       546,00 €
  • 129-3905
    Doypack® Paper/PET/CPP bags, 2×45 mm bottom gusset
    Doypack® Paper/PET/CPP bags, 2×45 mm bottom gusset
    1000 ml
    PET/CPP
    PAPER40/PET12/CPP60
    180×255 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 624,00 €
    624,00 €
    1 * 500 ST
       624,00 €