ContactUsLinkComponent
Glass Inserts for IM3 Microscopes
  OPTKIM3GLASS
 :  OPTIKA
Glass Inserts for IM3 Microscopes
  OPTKIM3GLASS
 :  OPTIKA
  • Description:
    Glass insert for IM3 microscope
  • Environmentally Preferable:
  • Pk:
    1
Inverted microscopes are useful for observing living cells or organisms at the bottom of a large container (e.g., a tissue culture flask) under more natural conditions than on a glass slide, as it occurs with a conventional microscope. IM-3 series is engineered and designed to be your ideal solution for fast and reliable routine inspections, with the exclusive, state-of-the-art X-LED8 illumination system.
A particularly simple and ingenious optical design allows stable alignments and smooth and accurate movements.
OPTIKA provides different configurations, including the innovative LED fluorescence technology for a new, enhanced
experience.