Minigrip® Antistatic Zip Seal Bags
Supplier: Flexico
Certificates
About this item
Minigrip® antistatic zip seal bags made up of PE.
- Average surface resistivity of ±1012 Ohms
- Half decay time of ±3 s
- Gives protection to the contents inside
Frequently Bought Together