Order Entry
Austria
ContactUsLinkComponent
Brady® SPC® Absorbents HAZWIK Quick Response Kit, Chemical
Brady® SPC® Absorbents HAZWIK Quick Response Kit, Chemical
Catalog # BRDY152186
Supplier:  Brady
undefined
Brady® SPC® Absorbents HAZWIK Quick Response Kit, Chemical
Catalog # BRDY152186
Supplier:  Brady

Specifications

  • Type:
    Chemical sorbent
  • Colour:
    Green
  • Description:
    HAZWIK quick response kit - chemical
  • W×D×H:
    228,6×482,6×333,5 mm
  • Pack type:
    BAGA
  • Absorbency:
    1,25 L
  • Pk:
    5

Specifications

About this item

Get fast and effective spill cleanup with a HAZWIK® quick response kit. It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.

  • Compact, single-use kit that is ideal for use on small spills in the laboratory
  • Quickly cleans up hazardous chemical spills
  • Visual instructions on the package
  • Contains PPE, tools, absorbent pads, acid and base neutralisers, wipes and disposable materials
Delivery: Includes 0,13 L acid neutraliser (1), 0,13 L base neutraliser (1), cable tie (1), disposal bag (1), pair of nitrile gloves (1), scoop and scraper (1), surface sanitising wipe (1), 190,5×254 mm pads (6).
Packaging: Packed as five individual kits per case.