Order Entry
Austria
ContactUsLinkComponent
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 with Ribbon for M510, BMP51, BMP53, M511 Printers
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 with Ribbon for M510, BMP51, BMP53, M511 Printers
Catalog # BRDY170928
Supplier:  Brady
undefined
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 with Ribbon for M510, BMP51, BMP53, M511 Printers
Catalog # BRDY170928
Supplier:  Brady

Specifications

  • Type:
    B-499
  • Colour:
    Black on white
  • Dimensions:
    38,01W mm × 4,87L m
  • Material:
    Nylon
  • Model:
    M5C-1500-499
  • Pk:
    1
  • Adhesive:
    Permanent acrylic

Specifications

About this item

Aggressive adhesive multi-purpose nylon labels is a tough and versatile label.

  • Versatile - Ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates and terminal blocks
  • Durable - Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Adhesive - Permanent adhesive labels that stick to smooth, rough and powder-coated surfaces alike

Application: Component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, wire and cable labelling.

Matte finish, abrasion-resistant, chemical-resistant, humidity-resistant, UV-resistant, weather-resistant, indoor use B-499 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.

Certifications: UL Recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant to 2011/65/EU (Pass 2015/863).