ContactUsLinkComponent
Sentinel® Back Harness
Sentinel® Back Harness
Catalog # ILCDS-3105
Supplier:  ILC DOVER
undefined
Sentinel® Back Harness
Catalog # ILCDS-3105
Supplier:  ILC DOVER
Specifications
  • Description:
    Back harness
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
The ergonomic back harness permits the Sentinel® blower to be worn comfortably on the back.
  • Ergonomic design
  • Blower securely snaps into the back harness
  • Supports longer shift operations
The harness features heat-sealed construction for easy cleaning, and is ideal for use throughout long shifts.