ContactUsLinkComponent
Sentinel® Flow Meter
  ILCDS-2010
 :  ILC DOVER
undefined
Sentinel® Flow Meter
  ILCDS-2010
 :  ILC DOVER
  • Description:
    Flow meter
  • Seq:
    04
  • Pk:
    1
Measures the air flow coming out of the blower to help ensure maximum protection.
  • Easy to use/read
  • Confirms proper flow rate is being achieved