ContactUsLinkComponent
96 Well Plate, RCH CEREX PWAX
96 Well Plate, RCH CEREX PWAX
Catalog # TCAT694-0201RCH
Supplier:  Tecan
96 Well Plate, RCH CEREX PWAX
Catalog # TCAT694-0201RCH
Supplier:  Tecan
Specifications
  • No. of wells:
    96
  • Description:
    96-well plate for RCH Cerex® PWAX
  • Phase:
    PWAX
  • Filling:
    20 mg
  • Pk:
    1
Specifications
About this item
A 18 µm polymer micro particulate with weak anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Packaging: 1 piece per box