Order Entry
Austria
ContactUsLinkComponent
Cerex® PSCX 96 Well Plate, RCH
Cerex® PSCX 96 Well Plate, RCH
Catalog # TCAT687-0101RCH
Supplier:  Tecan
Cerex® PSCX 96 Well Plate, RCH
Catalog # TCAT687-0101RCH
Supplier:  Tecan

Specifications

  • Description:
    Plate RCH Cerex PSCX, 1 ml, 25 µm
  • Phase:
    PSCX
  • Filling:
    10 mg
  • Pk:
    1

Specifications

About this item

A 25 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.

  • Cerex
  • Ion-exchange
  • Liquid handler compatible

Ideal for basic target compounds.

Packaging: 1 piece per box