Order Entry
Austria
ContactUsLinkComponent
Matte White Polypropylene RFID Laboratory Labels
Matte White Polypropylene RFID Laboratory Labels
Catalog # BRDY312403
Supplier:  Brady
undefined
Matte White Polypropylene RFID Laboratory Labels
Catalog # BRDY312403
Supplier:  Brady

Specifications

  • Colour:
    White
  • Description:
    RFID laboratory labels
  • Dimensions:
    65W×6H mm
  • Material:
    PP
  • Model:
    THT-65x6-7425/3004-0.5
  • Packed:
    500 lables per roll
  • Pk:
    1 Roll
  • Adhesive:
    Acrylic

Specifications

About this item

UHF RFID label for use on general laboratory identification on medical well plate, petri and cell culture dish.

  • Pharmaceutical and healthcare
  • The RFID label is designed for used on well plate, petri and cell culture dish in the laboratory environment
  • Polyester tag face material, with chip bonded to aluminium antenna using anisotropic conductive film adhesive, suitable for thermal transfer printing, including RFID encoding
  • Labels are outside wound and 100% inspected, 500 good RFID labels per roll with bad tags marked
Certifications: Compliant with directive 2011/65/EU and 2015/863.