Brady® FR22 RFID Fixed Reader
Product Details & Documents
The BRADY FR22 is the core device of a new approach for RFID fixed systems with wide communication options and a modular design. The FR22 is a critical building block for various track and trace and intelligent manufacturing solutions. It enables different use cases for different segments depending on which modules are attached to it.
- Fast and reliable edge computing with support for custom embedded applications
- New UHF RFID engine based on the very latest technology
- Brady extension connector for plug and play accessories like Brady MUX16
- Connect external devices, sensors, triggers, lights and buzzers
- Full set of API’s available for controlling the reader
Wide communication options that enable flexibility in system designing and no need for external connectivity hardware. Develop embedded web apps with user interface. Eliminating the need for external PC/tablet.
: ISO 18000-63 (EPC Class 1Gen2v2).
: Two years warranty (extendable), Free technical support during and after warranty. This box includes a Brady FR22 fixed reader, a WLAN/Bluetooth antenna, a safety and regulations card, additional product label with product information.
Specifications
- Description:FR22 fixed RFID reader
- Cat. No.:BRDY311937
- W×D×H:203,2×78,49×17,55 mm
- Power:100 - 240 V at 50 - 60 Hz
- Operating temperature:−20...+55 °C
Accessories
Product Information
- Catalog #AvailabilityPrice
- BFA868 RFID antenna for FR22 fixed RFID readerBFA868 RFID antenna for FR22 fixed RFID readerBRDY311942Log in for availability1 * 1 ST - 1 400,00 €
Product Information
- Catalog #AvailabilityPrice
- GA30 RFID antenna for FR22 fixed RFID reader
Product Information
- Catalog #AvailabilityPrice
- FR22 antenna cable 5 M SMA-MALE - RP-SMA
Product Information
- Catalog #AvailabilityPrice
- EU power supply for FR22 fixed RFID reader



