Order Entry
Austria
ContactUsLinkComponent
Brady® S3100 Sign and Label Printer F&B kit
Brady® S3100 Sign and Label Printer F&B kit
  BRDY311116
 :  Brady
undefined
Brady® S3100 Sign and Label Printer F&B kit
  BRDY311116
 :  Brady

 

  • Description:
    BradyPrinter S3100 sign and label printer - F&B kit, EU
  • Seq:
    01
  • Model:
    S3100
  • Print width:
    101,60 mm (max.)
  • Power supply:
    110 - 240 V
  • Connections:
    Ethernet, USB
  • Connector type:
    EU plug
  • Dimensions:
    304,80W×260,35D×241,30H mm
  • Display:
    Colour LCD touchscreen
  • Resolution:
    300 dpi
  • Weight:
    9,07 kg
  • Print method:
    Thermal transfer
  • Print speed:
    76,00 mm/s
  • Pk:
    1

 

 

The easy-to-use BradyPrinter S3100 sign and Label printer is designed to meet a variety of facility identification needs, helping to save time so we can get back to work.

  • •Easily design and print professional food or beverage safety signs, pipe markers and labels when you need them
  • •Print incredibly fast with automatic setup, built-in keyboard and touchscreen interface
  • •Create reliable labels that are washdown resistant and do not crumble when exposed to cleaning agents or pressure
  • •Fully protect food processing with metal detectable signs that enable additional protection against contamination
  • •Continuous and Die-cut Labels
  • •Monocolour colour capability

The built-in keyboard and touchscreen interface make label creation a breeze. Ideal for multiuser areas in your facility, anyone can simply walk up, create labels and print right on the spot.

Stand Alone Printing: Stand-alone and Peripheral
Max. Label Width (mm): 107,95 mm
Recommended Usage per Day: 600
Print Technology: Thermal Transfer

The S3100 label printer F&B kit used for component and equipment labelling, Exit and egress marking, facility identification, floor marking, general identification, hazard communication identification, lean and 5S labelling, pipe and valve marking, safety identification, Temperature indicating labelling.

Material supported: Magnetic Polyester, Metal-detectable Polyester, Metallic, Non-metal Polyester, Metallised Polyester, Paper, Phosphorescent Polyester, Polyester, Polyester with Polyester Overlaminate, Polyimide, Polyvinylfluoride, Reflective Tape, Tamper-evident Metallised Polyester, Vinyl, Vinyl Cloth

 : UL, cUL, CE, RoHS, RCM, EAC, CCC, China RoHS.
 : Includes: 1×white (B30C-4000-854-WT) and 1×Yellow (B30C-4000-854-YL), B30 Series R6000 Printer ribbon black (B30-R6000HF), Brady workstation basic design suite, S3100 signs and label printer (S3100-QY-UK), B30 Series ToughWash washdown resistant metal detectable labels.