ContactUsLinkComponent
Polyethylene Adhesion Tape
Polyethylene Adhesion Tape
Catalog # UTAP1110RE200-P3D
Supplier:  ULTRATAPE
Polyethylene Adhesion Tape
Catalog # UTAP1110RE200-P3D
Supplier:  ULTRATAPE
Specifications
  • Width:
    2"
  • Colour:
    Red
  • Description:
    Polyethylene adhesion tape
  • Environmentally Preferable:
  • Pk:
    3 Roll
Specifications
About this item
UltraTape 1110 is designed specifically for sealing wafer shipping boxes as well as cleanroom packaging and bag sealing. Its specially formulated medium-high adhesive leaves no residue or ghosting on plastic boxes, even after prolonged storage.