ContactUsLinkComponent
Microscope Polariser Analyser Set, L ICT/P for DM Series
Microscope Polariser Analyser Set, L ICT/P for DM Series
  LEIM11525359
Microscope Polariser Analyser Set, L ICT/P for DM Series
  LEIM11525359
  • Description:
    Analyser ICT/P
  • Seq:
    01
  • Pk:
    1
Leica microscope polariser analyser set L ICT/P for DM series.
  • Rotates 360°
  • Mounts on condenser bracket underneath condenser
  • Items are in good condition
  • These came from a Leica DMLB model