Order Entry
Austria
ContactUsLinkComponent
BT Lab Systems Horizontal Gel Electrophoresis Set
BT Lab Systems Horizontal Gel Electrophoresis Set
Catalog # GENOBT112
Supplier:  G-Biosciences
BT Lab Systems Horizontal Gel Electrophoresis Set
Catalog # GENOBT112
Supplier:  G-Biosciences

Specifications

  • Description:
    Horizontal gel electrophoresis set
  • Buffer volume:
    550 ml
  • Dimensions:
    300×160×75mm
  • Sample capacity:
    1 - 100 ml
  • Pk:
    1 SET

Specifications

About this item

BT Lab systems horizontal gel electrophoresis set's abundant buffer not only guarantees cooling effects, but also keeps the pH value stable during operation.

  • Contrasting sample background color on the tray for convenient sample loading
  • Auto-switch-off when the lid is removed

Gel sizes: 120×120 mm; 120×60 mm; 60×120 mm; 60×60 mm

Delivery: Combs supplied: 1/2/3 wells×1,5 mm, 6/13 wells×1 mm; 8/18 wells×1 mm; 11/25 wells×1 mm.