ContactUsLinkComponent
Handheld Wireless Palm Barcode Scanners: Bluetooth, 1D, 2D, QR Code, CR2700
Handheld Wireless Palm Barcode Scanners: Bluetooth, 1D, 2D, QR Code, CR2700
Supplier:  Brady
Certificates
undefined
Handheld Wireless Palm Barcode Scanners: Bluetooth, 1D, 2D, QR Code, CR2700
Supplier:  Brady
Specifications
  • Catalog No:
  • BRDY176517
  • BRDY176685
  • BRDY176686
  • BRDY176518
  • W×D×H:
  • 53,3×132,1×27,9 mm
  • 53,3×131,0×28,0 mm
  • 53,3×131,0×28,0 mm
  • 53,3×132,1×27,9 mm