Order Entry
Austria
ContactUsLinkComponent
 
SFC-100FLED Corded Stereo Microscope
Catalog # MOTI1100100800653
Supplier:  MOTIC
SFC-100FLED Corded Stereo Microscope
Catalog # MOTI1100100800653
Supplier:  MOTIC

Specifications

  • Model:
    SFC-100FLED corded
  • Description:
    Stereo microscope
  • Electrical:
    220 - 240 V (CE)
  • W×D×H:
    175×133×358 mm
  • Condenser:
    Built-in condenser lens N.A. 0,65
  • Observation method:
    Brightfield
  • Eyepieces:
    WF 10×/18 mm with pointer adjustment
  • Illumination:
    LED 3,5 V/70 mW
  • Nosepiece:
    Triple
  • Objectives:
    4× 0,10 (WD 15,9 mm), 10× 0,25 (WD 17,4 mm), 40× 0,65/S (WD 0,5 mm)
  • Optical design:
    Finite optical system, 160 mm
  • Stage:
    Plain stage with sample clips, 120×110 mm
  • Stand:
    Upright
  • Weight:
    2,7 kg
  • Pk:
    1
  • Type:
    Monocular

Specifications

About this item

The perfect entry into the world of microscopy; robust, easy to handle, and affordable: The Motic SFC100 Series offers the idea instrument for a first step into the 'world of small things'.

No plastics, but efficient optics 100% made of coated glass, this is the way to crisp and clear images for a long-term usage in classroom or for leisure activities. The SFC100 is a classic microscope with a classic performance.

Inclination: 45° inclined, 360° rotating
Objective classification: Achromatic, DIN
Objective mounting thread: W ⁴/₅×¹/₃₆ " (RMS standard)
Diaphragm: Iris diaphragm

Focus mechanism: Separate coarse and fine focusing adjustment with upper-limit stop to prevent damage to slides and objectives
Fine focus precision: 2 µm
Focusing stroke: 7 mm
Upper limit stop: Upper limit stop preset but adjustable

Filter: Built-in frosted glass
Transformer: Internal
Standard contrast technique: Brightfield

Accessories information: Dust cover included.