ContactUsLinkComponent
ratiolab® Pipette Tips, Colourless
ratiolab® Pipette Tips, Colourless
Catalog # RATI2400642
Supplier:  RATIOLAB
ratiolab® Pipette Tips, Colourless
Catalog # RATI2400642
Supplier:  RATIOLAB
Specifications
  • Sterile:
    -
  • Volume:
    1 - 10 ml
  • Tip colour:
    Clear
  • Pk:
    100 SET
Specifications
About this item
ratiolab® pipette tips are suitable for all commonly used pipette types.
  • Precision in each and every detail
  • Guarantee for highest quality
  • Made of high quality polypropylene
  • Autoclavable
  • Excellent transparency due to high quality surfaces