ContactUsLinkComponent
TELSTAR BiOptima EXA Biological Safety Cabinets, Class II
TELSTAR BiOptima EXA Biological Safety Cabinets, Class II
Certificates
undefined
TELSTAR BiOptima EXA Biological Safety Cabinets, Class II
Specifications
  • Catalog No:
  • TELR1738689
  • TELR1745204
  • Model:
  • BiOptima EXA 4
  • BiOptima EXA 6
  • Filters:
  • HEPA H14 (G3 prefilter)
  • HEPA H14 (G3 prefilter)
  • Noise level:
  • <52
  • <55
  • Electrical:
  • 230 V, 50 Hz, 200 W
  • 230 V, 50 Hz, 350 W
  • Ext. W×D×H:
  • 1338×797×2212 mm
  • 1948×797×2212 mm
  • Int. W×D×H:
  • 1190×548×666 mm
  • 1800×548×666 mm
  • Weight:
  • 285 kg
  • 380 kg