Order Entry
Austria
ContactUsLinkComponent
VWR® ConvoL NORMADOSE® Buffer Concentrate for Buffer Solution acc. to Weise for 1000 ml, pH 7.20
VWR® ConvoL NORMADOSE® Buffer Concentrate for Buffer Solution acc. to Weise for 1000 ml, pH 7.20
Catalog # 32075.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ConvoL NORMADOSE® Buffer Concentrate for Buffer Solution acc. to Weise for 1000 ml, pH 7.20
Catalog # 32075.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Reference temperature:
    20 °C
  • Description:
    ConvoL NORMADOSE® Buffer concentrate for buffer solution acc. to Weise
  • Final volume:
    1000 ml
  • pH value:
    7.20 ± 0.05

Specifications

About this item

This product is a ConvoL NORMADOSE® concentrate and contains exactly the amount of substance to prepare one liter of solution.