ContactUsLinkComponent
Bucket Liners
Bucket Liners
  MICRBKL993521IRB
 :  MICRONOVA
undefined
Bucket Liners
  MICRBKL993521IRB
 :  MICRONOVA
  • Description:
    Irradiated liner for B99 bucket
  • L×W:
    889×533,4 mm
  • Pk:
    24
The unique LDPE/nylon laminate is a superb barrier material for lining totes to transport raw materials or to line buckets containing disinfectants or other cleaning solutions.
  • Laminate is 75% LDPE, 25% nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts or raw material transfer
  • Thickness of 3 mil
  • Will hold up to autoclaving with liquid contents
  • Available sterile (irradiated) or non sterile
Non sterile liners are bulk packed. Select sterile liners are available bulk packed.
Fits: B-99 series buckets.
 : Flat packed, either individually double-bagged or in bulk packs. Packed in a case of 24 or case of 72 for bulk pack.