SymLine Lab Sinks with Drain and Mounting Sleeve
Lieferant: S.C.A.T.
Zertifikate
Über diesen Artikel
For integration in fume cupboards or on worktops for easy disposal of liquid solvent waste in large quantities.
- Made of dissipative PE-HD-EL
- Base with integrated slope for complete emptying
- Drain with removable sieve
- Laterally mounted earthing cable (length 1,5 m)
Secure mounting in the worktop is ensured by the fastening sleeves.
Dimension: 401×300×190 mm
Frequently Bought Together