Order Entry
Austria
ContactUsLinkComponent
Ätzend
Sulphuric acid 96% VLSI, Electronic Grade
Artikelnr. VWRC6682.2500
Lieferant:  VWR Chemicals
CAS-Nummer:  
undefined
Ätzend
Sulphuric acid 96% VLSI, Electronic Grade
Artikelnr. VWRC6682.2500
Lieferant:  VWR Chemicals
CAS-Nummer:  

Spezifikationen

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Spezifikationen

Spezifikation Testergebnisse

  • Assay (acidimetric):
    96.0 - 97.0 %
  • Colouration:
    ≤ 10 APHA
  • Particle size(>0.50 μm)(/ml)(at filling):
    ≤ 64
  • Cl (Chloride):
    ≤ 100 ppb
  • NH₄ (Ammonium):
    ≤ 500 ppb
  • NO₃ (Nitrate):
    ≤ 200 ppb
  • PO₄ (Phosphate):
    ≤ 200 ppb
  • Ag (Silver):
    ≤ 10 ppb
  • Al (Aluminium):
    ≤ 20 ppb
  • As + Sb (as As):
    ≤ 5 ppb
  • Au (Gold):
    ≤ 20 ppb
  • B (Boron):
    ≤ 10 ppb
  • Ba (Barium):
    ≤ 10 ppb
  • Be (Beryllium):
    ≤ 10 ppb
  • Bi (Bismuth):
    ≤ 20 ppb
  • Ca (Calcium):
    ≤ 100 ppb
  • Cd (Cadmium):
    ≤ 10 ppb
  • Co (Cobalt):
    ≤ 10 ppb
  • Cr (Chromium):
    ≤ 10 ppb
  • Cu (Copper):
    ≤ 10 ppb
  • Fe (Iron):
    ≤ 50 ppb
  • Ga (Gallium):
    ≤ 10 ppb
  • Ge (Germanium):
    ≤ 10 ppb
  • In (Indium):
    ≤ 10 ppb
  • K (Potassium):
    ≤ 50 ppb
  • Li (Lithium):
    ≤ 10 ppb
  • Mg (Magnesium):
    ≤ 20 ppb
  • Mn (Manganese):
    ≤ 10 ppb
  • Mo (Molybdenum):
    ≤ 10 ppb
  • Na (Sodium):
    ≤ 50 ppb
  • Ni (Nickel):
    ≤ 10 ppb
  • Pb (Lead):
    ≤ 10 ppb
  • Pt (Platinum):
    ≤ 20 ppb
  • Se (Selenium):
    ≤ 100 ppb
  • Sn (Tin):
    ≤ 20 ppb
  • Sr (Strontium):
    ≤ 10 ppb
  • Ti (Titanium):
    ≤ 10 ppb
  • Tl (Thallium):
    ≤ 20 ppb
  • V (Vanadium):
    ≤ 10 ppb
  • Zn (Zinc):
    ≤ 50 ppb
  • Zr (Zirconium):
    ≤ 10 ppb

Spezifikation Testergebnisse

Über diesen Artikel

 :
 :98.08 g/mol
Merck-Index:14,08974
Siedepunkt:330 °C (1013 hPa)
Schmelzpunkt:10.38 °C
Dichte:1.84 kg/l
Lagertemperatur:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
REACH:01-2119458838-20
 :GHS05
CAS-Nummer:7664-93-9
Formel:H₂SO₄
MDL-Nummer:MFCD00064589