Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 50% in aqueous solution, Selectipur®
Potassium hydroxide 50% in aqueous solution, Selectipur®
Catalog # 50488893.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydroxide 50% in aqueous solution, Selectipur®
Catalog # 50488893.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (acidimetric):
    49.5 - 50.5 %
  • Carbonate (as K₂CO₃):
    ≤ 0.5 %
  • Total N (Nitrogen):
    ≤ 5 ppm
  • Cl (Chloride):
    ≤ 25000 ppb
  • PO₄ (Phosphate):
    ≤ 5000 ppb
  • SO₄ (Sulphate):
    ≤ 10000 ppb
  • Al (Aluminium):
    ≤ 3000 ppb
  • As (Arsenic):
    ≤ 500000 ppb
  • Bi (Bismuth):
    ≤ 500000 ppb
  • Ca (Calcium):
    ≤ 3000 ppb
  • Cd (Cadmium):
    ≤ 500000 ppb
  • Co (Cobalt):
    ≤ 600 ppb
  • Cr (Chromium):
    ≤ 600 ppb
  • Cu (Copper):
    ≤ 300 ppb
  • Fe (Iron):
    ≤ 2000 ppb
  • Mg (Magnesium):
    ≤ 600 ppb
  • Mn (Manganese):
    ≤ 300 ppb
  • Na (Sodium):
    ≤ 3000000 ppb
  • Ni (Nickel):
    ≤ 300 ppb
  • Pb (Lead):
    ≤ 500000 ppb
  • Sb (Antimony):
    ≤ 500000 ppb
  • Sn (Tin):
    ≤ 500000 ppb
  • Zn (Zinc):
    ≤ 600 ppb

Specification Test Results

About this item

 :
 :56.11 g/mol
Density:1.475 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS07
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553