Ethylene glycol VLSI, Electronic Grade
Catalog # 50488857.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (GC):≥ 99.8 %
- Identification (IR):Passes test
- Miscibility with water:Passes test
- Colouration:≤ 10 APHA
- Particle size(>0.50 μm)(/ml)(at filling):≤ 2000
- Resistivity (MOhm-cm):≥ 1
- Evaporation residue:≤ 5 ppm
- Free acid (as CH₃COOH):≤ 10 ppm
- Free alkali (as NH₃):≤ 3 ppm
- Water:≤ 2000 ppm
- Cl (Chloride):≤ 4000 ppb
- Ag (Silver):≤ 20 ppb
- Al (Aluminium):≤ 100 ppb
- As (Arsenic):≤ 10 ppb
- Au (Gold):≤ 100 ppb
- B (Boron):≤ 10 ppb
- Ba (Barium):≤ 100 ppb
- Be (Beryllium):≤ 20 ppb
- Bi (Bismuth):≤ 100 ppb
- Ca (Calcium):≤ 100 ppb
- Cd (Cadmium):≤ 50 ppb
- Co (Cobalt):≤ 20 ppb
- Cr (Chromium):≤ 20 ppb
- Cu (Copper):≤ 20 ppb
- Fe (Iron):≤ 100 ppb
- Ga (Gallium):≤ 20 ppb
- In (Indium):≤ 20 ppb
- K (Potassium):≤ 100 ppb
- Li (Lithium):≤ 20 ppb
- Mg (Magnesium):≤ 100 ppb
- Mn (Manganese):≤ 20 ppb
- Mo (Molybdenum):≤ 50 ppb
- Na (Sodium):≤ 100 ppb
- Ni (Nickel):≤ 20 ppb
- Pb (Lead):≤ 50 ppb
- Pt (Platinum):≤ 200 ppb
- Sb (Antimony):≤ 10 ppb
- Sn (Tin):≤ 100 ppb
- Sr (Strontium):≤ 20 ppb
- Ti (Titanium):≤ 100 ppb
- Tl (Thallium):≤ 50 ppb
- V (Vanadium):≤ 50 ppb
- Zn (Zinc):≤ 100 ppb
- Zr (Zirconium):≤ 200 ppb
Specification Test Results
About this item
:
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:203-473-3
REACH:01-2119456816-28
:GHS07
CAS Number:107-21-1
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885