About this item
Air-Fed coverall with attached non-dssipative undergloves and integrated dissipative socks.
- Ventilated coverall with stitched and overtaped seams
- Fully enclosed over hood with clear front and crown see-through visor
- Front closure with double flap hook and loop, single zipper closing to center, horizontal opening
- Three exhaust valves; two at hips, one in hood back
- Air distribution made of Tychem 6000 fabric
- Emergency egress
- Attached non dissipative undergloves
- Integrated dissipative socks
Tychem® 6000 AL garments are made from a lightweight and durable fabric (<500 g per garment) consisting of a proprietary barrier film laminated to Tyvek® substrate. They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents.
Tychem® 6000 AL garments are designed to help keep workers safe and focused when carrying out critical tasks in industries like pharmaceutics, chemicals and others.
Colour: Grey
Attached under gloves:
Integrated dissipative socks with conductive Tyvek® tape strip.
Simplified closure: easier donning procedure, minimal help from an assistant required.
Airline belt inside the suit: high reusability of the belt/air-regulator.
Visor with 180° panoramic view: clear vision, non-PVC, highly chemical resistant solution.
Top visor: in suit lighting and vision above suit; improved safety with secured sights and comfort.
Emergency egress strip: quick doffing in case of emergency (tear off at once).
Fabric air plenum distributing cooling and breathing air for optimal comfort and reduced heat stress.
Simple breathing airline belt/regulator interface and installation: easy and quick installation set-up, no taping required.
Chemical protective clothing, Category III, Type 3-B, 4-B and 6-B
EN 14126 (barrier to infective agents)
EN 1073-1 (ventilated protective clothing against particulate radioactive contamination)
Antistatic treatment (EN 1149-5)
Certified system made of Tychem® 6000 AL TF630 WG and DuPont air-belt, model 8000513914, to protect the worker according to EN 14594 (Respiratory protective devices - Continuous flow compressed air line breathing devices)