Foam Nitrile Bamboo Gloves, NPR15
: Portwest
Extremely comfortable with excellent breathability, these gloves feature a 15-gauge liner constructed from bamboo viscose and nylon.
- Made using a sustainable bamboo liner
- Nitrile foam coating for excellent grip in wet and dry conditions
- Excellent durability and grip in wet and dry conditions
- 100% latex-free
- 15 gauge liner for extra dexterity
- Palm dipped to increase dexterity and ventilation
Bamboo is naturally anti-bacterial, hypoallergenic, and has resistance to UV light. The soft nitrile foam coated palm provides exceptional grip in wet and dry conditions. Perfect for a wide range of industries.
Colour: Black
Material: Bamboo/nylon/polyester
: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4121X. CE certified.
- Catalog No:
- 112-6605
- 112-6603
- 112-6604
- 112-6602
- 112-6600
- 112-6601
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
Frequently Bought Together