ContactUsLinkComponent
LPS for Endotoxin Indicator
LPS for Endotoxin Indicator
Catalog # WAKFWLPS-0100K
CAS Number:  
undefined
LPS for Endotoxin Indicator
Catalog # WAKFWLPS-0100K
CAS Number:  
Specifications
  • Pk:
    6
  • Description:
    LPS for Endotoxin indicator, with endotoxin level >100,000 EU/vial
Specifications
About this item
The LPS for Endotoxin Indicator is composed of E. coli O55:B5 (>100,000 EU/vial) and serves as a stock solution to prepare multiple endotoxin indicators for depyrogenation validation studies.
  • E. coli 055:B5 (>100,000 EU/vial)
  • Can make multiple endotoxin indicators from one vial
  • Used for dry heat oven validation studies
The prepared endotoxin indicators may be tested using reagents from our PYROSTAR™ ES-F series.