Order Entry
Austria
Orders LinkContactUsLinkComponent
Limulus PS Single Test

Limulus PS Single Test

LAL endotoxin-specific single-test vials, 20
Catalog # WAKF299-54501
Limulus PS Single Test
Catalog # WAKF299-54501
CAS Number:  
Limulus PS Single Test

LAL endotoxin-specific single-test vials, 20

CAS Number:  
Price
1 550,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

The Limulus PS Single Test is composed of LAL endotoxin-specific single-test vials and an affinity resin suspension called PyroSep™, designed to overcome any product interference by adsorbing potential endotoxin in samples while washing away the inhibitory components.

  • Endotoxin-specific reagent avoids false positive results from glucans
  • This kit is in single test configuration to avoid contamination
  • KTA quantitation for this product is performed on Toxinometer®
  • Allows endotoxin-specific measurement in samples that typically have inhibitory components: ethanol, silicone oil, soybean oil, olive oil, stearic acid, egg yolk lecithin, oil-adjuvanted vaccine, albumin products, coagulation factor products (factor 8), interferon formulation, antithrombin III, immunoglobulin product and other ethanol-soluble samples
  • Measurement system: Toxinometer

When using this product, the endotoxin in the sample is adsorbed onto a capillary column filled with PyroSep™ (an affinity adsorbent with histidine bound to a water-insoluble support via a spacer). Inhibitory substances contained in the sample are washed away, allowing the adsorbed endotoxin to be measured with the single-test format LAL reagent.

Specifications

Specifications

  • Pk:
    20
  • Description:
    LAL endotoxin-specific single-test vials

Specifications