Order Entry
Austria
ContactUsLinkComponent
XCORE UHPLC Columns, C18
XCORE UHPLC Columns, C18
Supplier:  ISERA
Certificates
XCORE UHPLC Columns, C18
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA411-A2CH00
  • ISERA411-A2CF00
  • ISERA411-A2CG00
  • ISERA411-A2CB00
  • ISERA411-A2CD00
  • ISERA411-A2CX00
  • ISERA411-A2CA00
  • ISERA411-A2CZ00
  • ISERA411-A2CU00
  • ISERA411-A2CV00
  • ISERA411-A2CK00
  • ISERA411-A2CM00
  • Phase name:
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • Endcapped:
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • Max. pressure:
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • USP code:
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Frequently Bought Together